missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dioxane, 98%

Manufacturer:  Alfa Aesar™ H5410706

 View more versions of this product

Catalog No. AAH5410706



Description

Description

Applications
1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

1,3-Dioxane
5°C (41°F)
105°C to 106°C
5g
C4H8O2
UN1165
1,3-dioxacyclohexane
VDFVNEFVBPFDSB-UHFFFAOYSA-N
1,3-dioxane
10450
88.11
1.032
1.418
-45°C
505-22-6
MFCD00006566
102532
Fully miscible in water.
C1COCOC1
88.106
CHEBI:46924
98%

Chemical Identifiers

505-22-6
88.106
VDFVNEFVBPFDSB-UHFFFAOYSA-N
10450
1,3-dioxane
C4H8O2
MFCD00006566
1,3-dioxacyclohexane
CHEBI:46924
C1COCOC1
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIOXANE

EINECSNumber : 208-005-1

RTECSNumber : JG8224000

TSCA : No

Recommended Storage : Ambient temperatures

Documents