missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Dimethylurea, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 116911000
Specifications
1, 3-Dimethylurea | |
Authentic | |
0.6% max. | |
White | |
100g | |
96-31-1 | |
100.0 | |
C3H8N2O | |
MFCD00008286 | |
1,3-dimethyl urea, dimethylurea, n,n'-dimethylharnstoff, n,n'-dimethylharnstoff german, n,n'-dimethylurea, sym-dimethylurea, symmetric dimethylurea, unii-wam6dr9i4x, urea, 1,3-dimethyl, urea, n,n'-dimethyl | |
MGJKQDOBUOMPEZ-UHFFFAOYSA-N | |
1,3-dimethylurea | |
7293 | |
88.11 | |
98% |
157°C | |
Plastic bottle | |
268°C to 270°C | |
101°C to 105°C | |
98% | |
97.5 | |
97.5% min. (HPLC) | |
CH3NHCONHCH3 | |
04, 65 | |
Solubility in water: 765g/L in water (21.5°C). Other solubilities: insoluble in ether,soluble in alcohols | |
CNC(=O)NC | |
88.11 | |
CHEBI:80472 | |
Crystals |
Chemical Identifiers
96-31-1 | |
88.11 | |
MGJKQDOBUOMPEZ-UHFFFAOYSA-N | |
7293 | |
1,3-dimethylurea |
C3H8N2O | |
MFCD00008286 | |
1,3-dimethyl urea, dimethylurea, n,n'-dimethylharnstoff, n,n'-dimethylharnstoff german, n,n'-dimethylurea, sym-dimethylurea, symmetric dimethylurea, unii-wam6dr9i4x, urea, 1,3-dimethyl, urea, n,n'-dimethyl | |
CHEBI:80472 | |
CNC(=O)NC |
Safety and Handling
EINECSNumber : 202-498-7
RTECSNumber : YS9868000
TSCA : TSCA