missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dimethylurea, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 116910010

 View more versions of this product

Catalog No. AC116910010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1, 3-Dimethylurea
Authentic
0.6% max.
White
1kg
96-31-1
100.0
C3H8N2O
MFCD00008286
1,3-dimethyl urea, dimethylurea, n,n'-dimethylharnstoff, n,n'-dimethylharnstoff german, n,n'-dimethylurea, sym-dimethylurea, symmetric dimethylurea, unii-wam6dr9i4x, urea, 1,3-dimethyl, urea, n,n'-dimethyl
MGJKQDOBUOMPEZ-UHFFFAOYSA-N
1,3-dimethylurea
7293
88.11
98%
157°C
Plastic bottle
268°C to 270°C
101°C to 105°C
98%
97.5
97.5% min. (HPLC)
CH3NHCONHCH3
04, 65
Solubility in water: 765g/L in water (21.5°C). Other solubilities: insoluble in ether,soluble in alcohols
CNC(=O)NC
88.11
CHEBI:80472
Crystals

Chemical Identifiers

96-31-1
88.11
MGJKQDOBUOMPEZ-UHFFFAOYSA-N
7293
1,3-dimethylurea
C3H8N2O
MFCD00008286
1,3-dimethyl urea, dimethylurea, n,n'-dimethylharnstoff, n,n'-dimethylharnstoff german, n,n'-dimethylurea, sym-dimethylurea, symmetric dimethylurea, unii-wam6dr9i4x, urea, 1,3-dimethyl, urea, n,n'-dimethyl
CHEBI:80472
CNC(=O)NC
Safety and Handling

Safety and Handling

EINECSNumber : 202-498-7

RTECSNumber : YS9868000

TSCA : TSCA

SDS Documents
Product Certifications