missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Dimethyluracil 98.0+%, TCI America™
$66.37 - $582.30
Chemical Identifiers
CAS | 874-14-6 |
---|---|
Molecular Formula | C6H8N2O2 |
Molecular Weight (g/mol) | 140.14 |
MDL Number | MFCD00038065 |
InChI Key | JSDBKAHWADVXFU-UHFFFAOYSA-N |
Synonym | 1,3-dimethyluracil, n1,n3-dimethyluracil, n,n'-dimethyluracil, 1,3-dimethylpyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 1,3-dimethyl, 2,4-dihydroxy-1,3-dimethylpyrimidine, uracil, 1,3-dimethyl, 1,3-dimethyl-2,4 1h,3h-pyrimidinedione, 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 1,3-dimethyl-1h-pyrimidine-2,4-dione |
PubChem CID | 70122 |
ChEBI | CHEBI:74763 |
IUPAC Name | 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione |
SMILES | CN1C=CC(=O)N(C)C1=O |
Chemical Identifiers
874-14-6 | |
140.14 | |
JSDBKAHWADVXFU-UHFFFAOYSA-N | |
70122 | |
1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione |
C6H8N2O2 | |
MFCD00038065 | |
1,3-dimethyluracil, n1,n3-dimethyluracil, n,n'-dimethyluracil, 1,3-dimethylpyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 1,3-dimethyl, 2,4-dihydroxy-1,3-dimethylpyrimidine, uracil, 1,3-dimethyl, 1,3-dimethyl-2,4 1h,3h-pyrimidinedione, 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 1,3-dimethyl-1h-pyrimidine-2,4-dione | |
CHEBI:74763 | |
CN1C=CC(=O)N(C)C1=O |
Specifications
874-14-6 | |
White | |
MFCD00038065 | |
1,3-dimethyluracil, n1,n3-dimethyluracil, n,n'-dimethyluracil, 1,3-dimethylpyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 1,3-dimethyl, 2,4-dihydroxy-1,3-dimethylpyrimidine, uracil, 1,3-dimethyl, 1,3-dimethyl-2,4 1h,3h-pyrimidinedione, 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 1,3-dimethyl-1h-pyrimidine-2,4-dione | |
CN1C=CC(=O)N(C)C1=O | |
140.14 | |
CHEBI:74763 | |
≥98.0% (GC) | |
1,3-Dimethyluracil |
125°C | |
C6H8N2O2 | |
1 g | |
JSDBKAHWADVXFU-UHFFFAOYSA-N | |
1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
70122 | |
140.14 | |
Crystalline Powder |
Safety and Handling
TSCA : Yes