Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dimethyluracil, 99%

$51.38 - $159.72

Chemical Identifiers

Chemical Name or Material 1,3-Dimethyluracil
CAS 874-14-6
Molecular Formula C6H8N2O2
Formula Weight 140.14
MDL Number MFCD00038065
Beilstein 124074
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1966403

  SDS
alfa aesar™
L1966403
1g Each for $51.38

AAL1966406

  SDS
alfa aesar™
L1966406
5g Each for $159.72
Description & Specifications

Specifications

Formula Weight 140.14
Chemical Name or Material 1,3-Dimethyluracil
Molecular Formula C6H8N2O2
Assay 99%
MDL Number MFCD00038065
Beilstein 124074
TSCA TSCA
Melting Point 119° to 125°C
Quantity 1g
CAS 874-14-6