missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dimethyluracil-5-carboxaldehyde, 96%

$99.28 - $341.72

Chemical Identifiers

CAS 4869-46-9
Molecular Formula C7H8N2O3
Formula Weight 168.15
MDL Number MFCD00099582
Synonym 5-Formyl-1,3-dimethyluracil
Chemical Name or Material 1,3-Dimethyluracil-5-carboxaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5028703 SDS Alfa Aesar™
H5028703
1g Each for $341.72
AAH50287MD SDS Alfa Aesar™
H50287MD
250mg Each for $99.28
Specifications Description & Specifications

Chemical Identifiers

4869-46-9
168.15
5-Formyl-1,3-dimethyluracil
C7H8N2O3
MFCD00099582
1,3-Dimethyluracil-5-carboxaldehyde

Specifications

4869-46-9
168.15
1g
1,3-Dimethyluracil-5-carboxaldehyde
C7H8N2O3
MFCD00099582
5-Formyl-1,3-dimethyluracil
96%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning