missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dimethylimidazolium methyl sulfate, 98%

$225.15 - $923.73

Chemical Identifiers

CAS 97345-90-9
Molecular Formula C6H12N2O4S
Molecular Weight (g/mol) 208.232
MDL Number MFCD03453276
InChI Key WOKQGMYCUGJNIJ-UHFFFAOYSA-M
Synonym 1,3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, 1,3-dimethyl-1h-imidazol-3-ium methyl sulfate, 1,3-dimethylimidazolium methyl sulfate 1-, dmimmeso4, 1, 3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, for catalysis hplc
PubChem CID 12075598
IUPAC Name 1,3-dimethylimidazol-1-ium;methyl sulfate
SMILES CN1C=C[N+](=C1)C.COS(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5992806 View Documents Alfa Aesar™
H5992806
5g Each for $225.15
 
AAH5992814 View Documents Alfa Aesar™
H5992814
25g Each for $923.73
 
Description

Description

Applications
It employed as a intermediate for pharmaceuticals.

Solubility
Soluble in water.

Notes
Moisture Sensitive, store away form water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent. Store under dry inert gas.
Specifications

Chemical Identifiers

97345-90-9
208.232
WOKQGMYCUGJNIJ-UHFFFAOYSA-M
12075598
CN1C=C[N+](=C1)C.COS(=O)(=O)[O-]
C6H12N2O4S
MFCD03453276
1,3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, 1,3-dimethyl-1h-imidazol-3-ium methyl sulfate, 1,3-dimethylimidazolium methyl sulfate 1-, dmimmeso4, 1, 3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, for catalysis hplc
1,3-dimethylimidazol-1-ium;methyl sulfate

Specifications

5g
C6H12N2O4S
9352735
WOKQGMYCUGJNIJ-UHFFFAOYSA-M
1,3-dimethylimidazol-1-ium;methyl sulfate
12075598
98%
97345-90-9
MFCD03453276
1,3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, 1,3-dimethyl-1h-imidazol-3-ium methyl sulfate, 1,3-dimethylimidazolium methyl sulfate 1-, dmimmeso4, 1, 3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, for catalysis hplc
CN1C=C[N+](=C1)C.COS(=O)(=O)[O-]
208.232
208.24
1,3-Dimethylimidazolium methyl sulfate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning