Please login to your online account to display your discounted pricing

1,3-Dimethylimidazolidin-2-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1, 3-Dimethylimidazolidin-2-one
CAS 80-73-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H10N2O
Formula Weight 114.15
Synonym DMEU
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01386FA

  SDS
fisher scientific company
SB01386FA
Amber Glass Bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.15
Quantity 100g
Packaging Amber Glass Bottle
Assay 97%
Density 1.0400g/mL
CAS Min % 97.0
Synonym DMEU
Assay Percent Range 97%
Molecular Formula C5H10N2O
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 80-73-9
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Chemical Name or Material 1, 3-Dimethylimidazolidin-2-one
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.