Please login to your online account to display your discounted pricing

1,3-Dimethylcyclohexane, 97%, mixture of cis and trans, Acros Organics

$21.00 - $21.00

Chemical Identifiers

Chemical Name or Material 1, 3-Dimethylcyclohexane
Name Note 97%
CAS 591-21-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H16
Linear Formula C6H10(CH3)
Formula Weight 112.21
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC338230050

  SDS
thermo fisher scientific
33823-0050
5mL Each for $21.00

AC338230250

  SDS
thermo fisher scientific
33823-0250
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 112.21
Physical Form Liquid
Quantity 5mL
Color Clear colorless
EINECS Number 209-707-0
TSCA TSCA
Density 0.77g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C8H16
Refractive Index 1.4260 to 1.4310 (20°C, 589nm)
Chemical Name or Material 1, 3-Dimethylcyclohexane
Linear Formula C6H10(CH3)
Assay Percent Range 97%
CAS 591-21-9
Name Note 97%