Please login to your online account to display your discounted pricing

1-(3-Dimethylaminopropyl)piperazine 97% , ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391070050

 View more versions of this product

Catalog No. AC391070050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 171.29
Physical Form Liquid
Quantity 5g
Color Clear light yellow
Density 0.9170g/mL
CAS Min % 96.0
CAS Max % 100.0
CAS 877-96-3
Infrared Spectrum Authentic
Chemical Name or Material 1-(3-Dimethylaminopropyl)piperazine
Assay Percent Range 97%
Molecular Formula C9H21N3
MDL Number MFCD00082601
Refractive Index 1.4770 to 1.4790 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-(3-Dimethylaminopropyl)piperazine
CAS 877-96-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H21N3
Formula Weight 171.29
MDL Number MFCD00082601
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents