missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America D1601250G

 View more versions of this product

Catalog No. D1601250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide Hydrochloride [Coupling Agent for Peptides Synthesis]
115°C
6.0 - 8.0 (50g/L)
MFCD00012503
1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl
Cl.CCN=C=NCCCN(C)C
191.70
191.70
Crystalline Powder
25952-53-8
White
C8H18ClN3
250 g
FPQQSJJWHUJYPU-UHFFFAOYSA-N
({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
2723939
≥98.0% (T)

Chemical Identifiers

25952-53-8
191.70
FPQQSJJWHUJYPU-UHFFFAOYSA-N
2723939
Cl.CCN=C=NCCCN(C)C
C8H18ClN3
MFCD00012503
1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl
({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1696

RTECSNumber : FF2200000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions