missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2505706

 View more versions of this product

Catalog No. AAB2505706



Description

Description

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide is used as a carboxyl activating agent and activate phosphate groups in phospho mono and di esters. It is used in peptide synthesis, 3'-amino-3'-deoxyadenosine-5'-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide is used as a carboxyl activating agent and activate phosphate groups in phospho mono and di esters. It is used in peptide synthesis, 3′-amino-3′-deoxyadenosine-5′-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with strong acids and strong oxidizing agents.
Specifications

Specifications

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide
0.885
>110°C (230°F)
1.461
MFCD00044916
UN2735
Air and moisture sensitive
Soluble in water.
CCN=C=NCCCN(C)C
155.245
155.24
1892-57-5
66°C to 68°C (1 mmHg)
C8H17N3
CH3CH2N=C=N(CH2)3N(CH3)2
5 g
507429
1-3-dimethylaminopropyl-3-ethylcarbodiimide, edac, 1-ethyl-3-3-dimethylaminopropyl carbodiimide, n1-ethylimino methylene-n3,n3-dimethylpropane-1,3-diamine, unii-rj5ozg6i4a, edci, 1,3-propanediamine, n'-ethylcarbonimidoyl-n,n-dimethyl, n-3-dimethylaminopropyl-n'-ethylcarbodiimide, rj5ozg6i4a, ethyldimethylaminopropyl carbodiimide
LMDZBCPBFSXMTL-UHFFFAOYSA-N
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine
15908
97%

Chemical Identifiers

1892-57-5
155.245
LMDZBCPBFSXMTL-UHFFFAOYSA-N
15908
CCN=C=NCCCN(C)C
C8H17N3
MFCD00044916
1-3-dimethylaminopropyl-3-ethylcarbodiimide, edac, 1-ethyl-3-3-dimethylaminopropyl carbodiimide, n1-ethylimino methylene-n3,n3-dimethylpropane-1,3-diamine, unii-rj5ozg6i4a, edci, 1,3-propanediamine, n'-ethylcarbonimidoyl-n,n-dimethyl, n-3-dimethylaminopropyl-n'-ethylcarbodiimide, rj5ozg6i4a, ethyldimethylaminopropyl carbodiimide
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 217-579-2

TSCA : Yes

Recommended Storage : Store at -20°C; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only