Please login to your online account to display your discounted pricing

1,3-Dimethylaminoadamantane Hydrochloride 99%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC298080500

 View more versions of this product

Catalog No. AC298080500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Powder
Assay ≥98.5% (total base)
Formula Weight 215.77
Quantity 50g
Color White
Assay Percent Range 99%
Chemical Name or Material 1, 3-Dimethyl-5-aminoadamantane hydrochloride
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C12H21N·HCl
CAS 41100-52-1

Specifications

Chemical Name or Material 1, 3-Dimethyl-5-aminoadamantane hydrochloride
Assay ≥98.5% (total base)
Assay Percent Range 99%
Molecular Formula C12H21N·HCl
Formula Weight 215.77