Please login to your online account to display your discounted pricing

1,3-dimethyl Uracil 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC381690010

 View more versions of this product

Catalog No. AC381690010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 140.14
Physical Form Needles
Quantity 1g
Color White to light yellow
Melting Point 119° to 124°C
CAS Max % 100.0
Synonym 1, 3-Dimethyl-2, 4(1H
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 874-14-6
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Chemical Name or Material 1, 3-Dimethyl uracil
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C6 H8 N2 O2
Assay Percent Range 98%
MDL Number MFCD00038065

Specifications

Chemical Name or Material 1, 3-Dimethyl uracil
CAS 874-14-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6 H8 N2 O2
Formula Weight 140.14
MDL Number MFCD00038065
Synonym 1, 3-Dimethyl-2, 4(1H
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents