Please login to your online account to display your discounted pricing

1,3-dimethyl Uracil 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Dimethyl uracil
CAS 874-14-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6 H8 N2 O2
Formula Weight 140.14
MDL Number MFCD00038065
Synonym 1, 3-Dimethyl-2, 4(1H
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381690010

  SDS
thermo fisher scientific
AC381690010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Needles
Formula Weight 140.14
Quantity 1g
Color White to light yellow
Melting Point 119° to 124°C
CAS Max % 100.0
CAS 874-14-6
Synonym 1, 3-Dimethyl-2, 4(1H
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1, 3-Dimethyl uracil
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C6 H8 N2 O2
MDL Number MFCD00038065
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.