Please login to your online account to display your discounted pricing

1,3-Dimethyl-5-morpholino-1H-pyrazol-4-amine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC19214DA

 View more versions of this product

Catalog No. CC19214DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 196.25
Quantity 1g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
MDL Number MFCD04115380
Molecular Formula C9H16N4O
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
CAS Min % 96.5
Packaging Amber Glass Bottle
Synonym 1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine (IUPAC)
Chemical Name or Material 1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 568577-87-7

Specifications

Chemical Name or Material 1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine
CAS 568577-87-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H16N4O
Formula Weight 196.25
MDL Number MFCD04115380
Synonym 1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine (IUPAC)
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents