missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dimethyl-5-morpholino-1H-pyrazol-4-amine, 97%, Maybridge

Chemical Identifiers

CAS 568577-87-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H16N4O
Formula Weight 196.25
MDL Number MFCD04115380
Synonym 1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine (IUPAC)
Chemical Name or Material 1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC19214DA SDS Fisher Scientific Company
CC19214DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

568577-87-7
100.0
C9H16N4O
MFCD04115380
1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine
96.5
97%
196.25
1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine (IUPAC)
97%

Specifications

196.25
100.0
MFCD04115380
96.5
1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine (IUPAC)
1, 3-Dimethyl-5-morpholino-1H-pyrazol-4-amine
1g
97%
C9H16N4O
Amber Glass Bottle
568577-87-7
97%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.