missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 97%, pure, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 224645000

 View more versions of this product

Catalog No. AC224645000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
128.17
500mL
1.064g/mL
11,207; 13,122; 16,146
100.0
Authentic
1.4875 to 1.4895 (20°C, 589nm)
97%
C6H12N2O
MFCD00006550
Extra Pure
Liquid
Clear colorless to slightly yellow
24, IV, 32
96.0
DMPU
Glass bottle
1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone
7226-23-5
97%

Chemical Identifiers

7226-23-5
100.0
C6H12N2O
MFCD00006550
1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 97%
Extra Pure
96.0
97%
128.17
DMPU
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Suspected of damaging fertility.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 230-625-6

RTECSNumber : UW7582500

TSCA : TSCA