missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dimethyl-2-thiohydantoin, 95%

Manufacturer:  Alfa Aesar™ H6338406

 View more versions of this product

Catalog No. AAH6338406



Specifications

Specifications

1,3-Dimethyl-2-thiohydantoin
5g
C5H8N2OS
1,3-dimethyl-2-thiohydantoin, 1,3-dimethyl-2-thioxoimidazolidin-4-one, n,n'-dimethylthiohydantoin, 1,3-dimethyl-2-thioxo-4-imidazolidinone, 4-imidazolidinone,1,3-dimethyl-2-thioxo, acmc-1c9l4, 1,3-dimethyl-2-thioxo-1,3-diazolidin-4-one
CN1CC(=O)N(C1=S)C
144.192
144.19
95%
Orange
1801-62-3
MFCD00186242
GKWNLFPHQCFFQJ-UHFFFAOYSA-N
1,3-dimethyl-2-sulfanylideneimidazolidin-4-one
3722203
Solid

Chemical Identifiers

1801-62-3
144.192
GKWNLFPHQCFFQJ-UHFFFAOYSA-N
3722203
CN1CC(=O)N(C1=S)C
C5H8N2OS
MFCD00186242
1,3-dimethyl-2-thiohydantoin, 1,3-dimethyl-2-thioxoimidazolidin-4-one, n,n'-dimethylthiohydantoin, 1,3-dimethyl-2-thioxo-4-imidazolidinone, 4-imidazolidinone,1,3-dimethyl-2-thioxo, acmc-1c9l4, 1,3-dimethyl-2-thioxo-1,3-diazolidin-4-one
1,3-dimethyl-2-sulfanylideneimidazolidin-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents