Please login to your online account to display your discounted pricing

1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid, 95+%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC22901DA

 View more versions of this product

Catalog No. CC22901DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 196.23
Quantity 1g
Assay 95%
Chemical Name or Material 1, 3-Dimethyl-1H-thieno[2, 3-c]pyrazole-5-carboxylic acid
Assay Percent Range 95%
Molecular Formula C8H8N2O2S
Packaging Amber Glass Bottle
CAS 25252-46-4

Specifications

Chemical Name or Material 1, 3-Dimethyl-1H-thieno[2, 3-c]pyrazole-5-carboxylic acid
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H8N2O2S
Formula Weight 196.23