1,3-Dimethyl-1H-pyrazole-5-carboxylic acid, 97%, Acros Organics™
Manufacturer:  Acros Organics AC448630100
 View more versions of this product
Catalog No. AC448630100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
209.0°C to 212.0°C
5744-56-9
1,3-dimethyl-1h-pyrazole-5-carboxylic acid, 1,3-dimethylpyrazole-5-carboxylic acid, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl, 1,3-dimethylpyrazole-5-carboxylicacid, pubchem7839, ksc269g2l, 1,3-dimethylpyrazol-5-carboxylic acid, pyrazole-5-carboxylic acid, 1,3-dmethyl, 1h-pyrazole-5-carboxylicacid, 1,3-dimethyl
CC1=NN(C(=C1)C(=O)O)C
140.142
140.14
Authentic
10g
C6H8N2O2
QRWZFUBHOQWUGH-UHFFFAOYSA-N
2,5-dimethylpyrazole-3-carboxylic acid
587721
97%
Chemical Identifiers
5744-56-9
140.142
1,3-dimethyl-1h-pyrazole-5-carboxylic acid, 1,3-dimethylpyrazole-5-carboxylic acid, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl, 1,3-dimethylpyrazole-5-carboxylicacid, pubchem7839, ksc269g2l, 1,3-dimethylpyrazol-5-carboxylic acid, pyrazole-5-carboxylic acid, 1,3-dmethyl, 1h-pyrazole-5-carboxylicacid, 1,3-dimethyl
2,5-dimethylpyrazole-3-carboxylic acid
C6H8N2O2
QRWZFUBHOQWUGH-UHFFFAOYSA-N
587721
CC1=NN(C(=C1)C(=O)O)C
Safety and Handling
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Warning
SDS Documents