Please login to your online account to display your discounted pricing

1:3-Dimethyl-1H-pyrazole-5-carbothioamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carbothioamide
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H9N3S
Formula Weight 155.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC12518CB

  SDS
fisher scientific company
CC12518CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC12518DA

  SDS
fisher scientific company
CC12518DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC12518DE

 
fisher scientific company
CC12518DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.22
Quantity 250mg
Assay 97%
Molecular Formula C6H9N3S
Assay Percent Range 97%
Packaging Amber Glass Bottle
Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carbothioamide
CAS 844891-03-8