Please login to your online account to display your discounted pricing

1:3-Dimethyl-1H-pyrazole-5-carbothioamide, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC12518DE

 View more versions of this product

Catalog No. CC12518DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 155.22
Quantity 5g
Assay 97%
Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carbothioamide
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C6H9N3S
CAS 844891-03-8

Specifications

Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carbothioamide
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H9N3S
Formula Weight 155.22