Please login to your online account to display your discounted pricing

1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride, Maybridge

Chemical Identifiers

Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7ClN2O
Formula Weight 158.59
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC12502EA

 
fisher scientific company
CC12502EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC12502CB

 
fisher scientific company
CC12502CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.59
Quantity 10g
Assay 97%
Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Molecular Formula C6H7ClN2O
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 55458-67-8