Please login to your online account to display your discounted pricing

1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC12502CB

 View more versions of this product

Catalog No. CC12502CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 158.59
Quantity 250mg
Assay 97%
Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 55458-67-8
Molecular Formula C6H7ClN2O

Specifications

Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7ClN2O
Formula Weight 158.59