Please login to your online account to display your discounted pricing

(1,3-Dimethyl-1H-pyrazol-5-yl)methylamine, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC12513DE

 View more versions of this product

Catalog No. CC12513DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 125.17
Quantity 5g
Assay 95%
Molecular Formula C6H11N3
CAS 499770-63-7
Chemical Name or Material (1, 3-Dimethyl-1H-pyrazol-5-yl)methylamine
Packaging Amber Glass Bottle
Assay Percent Range 95%

Specifications

Chemical Name or Material (1, 3-Dimethyl-1H-pyrazol-5-yl)methylamine
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H11N3
Formula Weight 125.17