Please login to your online account to display your discounted pricing

(1,3-Dimethyl-1H-pyrazol-5-yl)methylamine, 97%, Maybridge

$71.36 - $71.36

Chemical Identifiers

Chemical Name or Material (1, 3-Dimethyl-1H-pyrazol-5-yl)methylamine
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H11N3
Formula Weight 125.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC12513DE

 
fisher scientific company
CC12513DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC12513DA

  SDS
fisher scientific company
CC12513DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC12513CB

  SDS
fisher scientific uk ltd
CC12513CB
Amber Glass Bottle 250mg Each for $71.36
Description & Specifications

Specifications

Formula Weight 125.17
Quantity 5g
Assay 95%
Molecular Formula C6H11N3
CAS 499770-63-7
Chemical Name or Material (1, 3-Dimethyl-1H-pyrazol-5-yl)methylamine
Packaging Amber Glass Bottle
Assay Percent Range 95%