Please login to your online account to display your discounted pricing

(1,3-Dimethyl-1H-pyrazol-5-yl)methylamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific uk ltd  CC12513CB

 View more versions of this product

Catalog No. CC12513CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 125.17
Quantity 250mg
Assay 95%
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C6H11N3
Chemical Name or Material (1, 3-Dimethyl-1H-pyrazol-5-yl)methylamine
CAS 499770-63-7

Specifications

Chemical Name or Material (1, 3-Dimethyl-1H-pyrazol-5-yl)methylamine
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H11N3
Formula Weight 125.17