missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Diisopropylimidazolium chloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 139143-09-2
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H17ClN2
Formula Weight 188.7
MDL Number MFCD03840577
Chemical Name or Material 1, 3-Diisopropylimidazolium chloride
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC382430050 SDS Thermo Fisher Scientific
AC382430050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

139143-09-2
100.0
C9H17ClN2
MFCD03840577
≥96.0%
96.0
97%
188.7
1, 3-Diisopropylimidazolium chloride

Specifications

Crystalline Powder
5g
96.0
Authentic
MFCD03840577
Glass bottle
1, 3-Diisopropylimidazolium chloride
188.7
Off-white to light beige
100.0
C9H17ClN2
97%
139143-09-2
≥96.0%