Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Diisopropylimidazolium chloride, 97+%

$68.81 - $240.12

Chemical Identifiers

Chemical Name or Material 1,3-Diisopropylimidazolium chloride
CAS 139143-09-2
Molecular Formula C9H17ClN2
Formula Weight 188.69
MDL Number MFCD03840577
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2779803

  SDS
alfa aesar™
H2779803
1g Each for $68.81

AAH2779806

  SDS
alfa aesar™
H2779806
5g Each for $240.12
Description & Specifications

Specifications

Formula Weight 188.69
Chemical Name or Material 1,3-Diisopropylimidazolium chloride
Melting Point 182° to 186°C
Assay ≥97%
Molecular Formula C9H17ClN2
Quantity 1g
MDL Number MFCD03840577
CAS 139143-09-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Ligand for ruthenium-catalyzed greener amide bond formation by dehydrogenative coupling of amines and alcohols.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.