missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Diiodopropane, 99%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 218601000

 View more versions of this product

Catalog No. AC218601000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,3-Diiodopropane
627-31-6
100.0
2.5760g/mL
>110°C
98.5% min. (GC)
C3H6I2
I(CH2)3I
100 mL
2.576
Solubility in water: insoluble in water.
C(CI)CI
295.88
12314
99%
99%
98.5
Pink to Yellow
111.0°C to 113.0°C (31.0 mmHg)
Authentic
Glass bottle
1.6405 to 1.6425
MFCD00001097
01, 115
trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer
AAAXMNYUNVCMCJ-UHFFFAOYSA-N
1,3-diiodopropane
copper
295.88
Liquid

Chemical Identifiers

627-31-6
295.88
AAAXMNYUNVCMCJ-UHFFFAOYSA-N
12314
C(CI)CI
C3H6I2
MFCD00001097
trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer
1,3-diiodopropane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-993-4

RTECSNumber : TY1080000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only