Learn More
1,3-Diiodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1061618
Description
1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Stable, but light sensitive - store in the dark. Incompatible with strong bases, strong oxidizing agents.
Specifications
1,3-Diiodopropane | |
627-31-6 | |
2.565 | |
>110°C (230°F) | |
1.6415 | |
50 g | |
Light sensitive | |
Insoluble in water. | |
C(CI)CI | |
295.89 | |
295.89 |
Stabilized with copper | |
-20°C | |
224°C | |
C3H6I2 | |
MFCD00001097 | |
1731117 | |
trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer | |
AAAXMNYUNVCMCJ-UHFFFAOYSA-N | |
1,3-diiodopropane | |
12314 | |
≥98% |
Chemical Identifiers
627-31-6 | |
295.89 | |
AAAXMNYUNVCMCJ-UHFFFAOYSA-N | |
12314 | |
C(CI)CI |
C3H6I2 | |
MFCD00001097 | |
trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer | |
1,3-diiodopropane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-993-4
RTECSNumber : TY1080000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only