missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dihydroxynaphthalene, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163340250

 View more versions of this product

Catalog No. AC163340250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 3-Dihydroxynaphthalene
Glass bottle
123.0°C to 126.0°C
132-86-5
C10H8O2
06,978
1,3-dihydroxynaphthalene
XOOMNEFVDUTJPP-UHFFFAOYSA-N
naphthalene-1,3-diol
8601
Crystalline Powder
Authentic
Brown to Orange-Pink
25g
99+%
C10H6(OH)2
15,6481
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether,soluble in ethanol an diethyl ether,slightly soluble in acetone,benzene and ligroin
C1=CC=C2C(=C1)C=C(C=C2O)O
160.17
160.17
99+%

Chemical Identifiers

132-86-5
160.17
1,3-dihydroxynaphthalene
naphthalene-1,3-diol
C10H8O2
XOOMNEFVDUTJPP-UHFFFAOYSA-N
8601
C1=CC=C2C(=C1)C=C(C=C2O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 205-079-7

SDS Documents