Please login to your online account to display your discounted pricing

1,3-Dihydro-1-(1-methylethenyl)-2H-benzimidazole-2-one, 99%, ACROS Organics™

$181.47 - $181.47

Chemical Identifiers

Chemical Name or Material 1-Isopropenyl-2-benzimidazolidinone
CAS 52099-72-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥99.6%
Assay Percent Range 99%
Molecular Formula C10 H10 N2 O
Formula Weight 174.2
MDL Number MFCD00218253
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC100290050

  SDS
thermo fisher scientific
10029-0050
Glass bottle 5g Each for $181.47
Description & Specifications

Specifications

Assay ≥99.6%
Formula Weight 174.2
Physical Form Crystalline Powder
Quantity 5g
Color White to slightly cream
Melting Point 120° to 124°C
CAS Max % 100.0
Chemical Name or Material 1-Isopropenyl-2-benzimidazolidinone
Packaging Glass bottle
Total Impurity < 0.2% (HPLC)
Assay Percent Range 99%
MDL Number MFCD00218253
CAS 52099-72-6
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Individual Impurity < 0.1% (HPLC)
2_Hydroxybenzimidazole 2-hydroxybenzimidazole (HPLC): < 0.2%
Infrared Spectrum Authentic
Molecular Formula C10 H10 N2 O
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.