Please login to your online account to display your discounted pricing

1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  135910010

 View more versions of this product

Catalog No. AC135910010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0% (HPLC)
Physical Form Powder
Formula Weight 215.25
Quantity 1g
Color Light beige to beige
Water 5% Max. (K.F.)
Melting Point 195° to 200°C
Chemical Name or Material 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Assay Percent Range 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C12H13N3O
CAS 2147-83-3
Lambda Max 280 to 284nm

Specifications

Chemical Name or Material 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C12H13N3O
Formula Weight 215.25