Please login to your online account to display your discounted pricing

1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one, 97%, ACROS Organics™

$75.15 - $151.49

Chemical Identifiers

Chemical Name or Material 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C12H13N3O
Formula Weight 215.25
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC135910010

  SDS
thermo fisher scientific
135910010
Glass bottle 1g Each for $151.49

AC135912500

  SDS
thermo fisher scientific
135912500
Each for $75.15

AC135910050

  SDS
thermo fisher scientific
AC135910050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0% (HPLC)
Physical Form Powder
Formula Weight 215.25
Quantity 1g
Color Light beige to beige
Water 5% Max. (K.F.)
Melting Point 195° to 200°C
Chemical Name or Material 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Assay Percent Range 97%
Infrared Spectrum Authentic
Molecular Formula C12H13N3O
Packaging Glass bottle
Lambda Max 280 to 284nm
CAS 2147-83-3