Please login to your online account to display your discounted pricing

1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC135910050

 View more versions of this product

Catalog No. AC135910050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Quantity 5g
CAS 2147-83-3
Lambda Max 280 to 284nm
Infrared Spectrum Authentic
Packaging Glass bottle
Water 5% Max. (K.F.)
Physical Form Powder
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C12H13N3O
Color Light beige to beige
Formula Weight 215.25
Melting Point 195° to 200°C

Specifications

Chemical Name or Material 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C12H13N3O
Formula Weight 215.25