Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Difluoropropane, 97%

$61.03 - $253.14

Chemical Identifiers

Chemical Name or Material 1,3-Difluoropropane
CAS 462-39-5
Molecular Formula C3H6F2
Linear Formula FCH2CH2CH2F
Formula Weight 80.08
MDL Number MFCD00040969
Beilstein 1696999
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1235106

  SDS
alfa aesar™
L1235106
5g Each for $61.03

AAL1235114

  SDS
alfa aesar™
L1235114
25g Each for $253.14
Description & Specifications

Specifications

Formula Weight 80.08
Chemical Name or Material 1,3-Difluoropropane
Molecular Formula C3H6F2
Assay 97%
CAS 462-39-5
Quantity 5g
Linear Formula FCH2CH2CH2F
Beilstein 1696999
Recommended Storage 2° to 8°C
Boiling Point 40° to 42°C
MDL Number MFCD00040969
Safety and Handling

2° to 8°C