Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Difluorobenzene, 99%

$73.74 - $73.74

Chemical Identifiers

Chemical Name or Material 1,3-Difluorobenzene
CAS 372-18-9
Molecular Formula C6H4F2
Formula Weight 114.09
MDL Number MFCD00000327
UN Number UN1993
Beilstein 1904537
Synonym m-Difluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1438709

  SDS
alfa aesar™
A1438709*
10g Each for $73.74

AAA1438718

  SDS
alfa aesar™
A1438718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1438730

  SDS
alfa aesar™
A1438730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.09
Chemical Name or Material 1,3-Difluorobenzene
Assay 99%
Molecular Formula C6H4F2
MDL Number MFCD00000327
CAS 372-18-9
UN Number UN1993
Beilstein 1904537
Synonym m-Difluorobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P260-P243-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number CZ5652000
Flash Point 2°C (36°F)
Odor Characteristic
Boiling Point 82° to 83°C
Melting Point −59°C
Quantity 10g

Applications
It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.

Solubility
Soluble in water at 20°C 1.1g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Danger

GHS P Statement
P210-P260-P243-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.