missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Difluoro-5-nitrobenzene, 98%

$34.70 - $139.70

Chemical Identifiers

CAS 2265-94-3
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.092
MDL Number MFCD00012142
InChI Key AUQBBDWDLJSKMI-UHFFFAOYSA-N
Synonym 3,5-difluoronitrobenzene, benzene, 1,3-difluoro-5-nitro, 1,3-difluoro-5-nitro-benzene, 3,5-difluoro-1-nitrobenzene, 1-nitro-3,5-difluorobenzene, benzene, 1,3-difluoro-5-nitro-7ci,8ci,9ci, 3,5-difluoronitroben, pubchem2291, 3,5-difluornitrobenzene, acmc-1ck2x
PubChem CID 75272
IUPAC Name 1,3-difluoro-5-nitrobenzene
SMILES C1=C(C=C(C=C1F)F)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6027003 View Documents Alfa Aesar™
H6027003
1g Each for $34.70
 
AAH6027006 View Documents Alfa Aesar™
H6027006
5g Each for $139.70
 
Specifications

Chemical Identifiers

2265-94-3
159.092
AUQBBDWDLJSKMI-UHFFFAOYSA-N
75272
C1=C(C=C(C=C1F)F)[N+](=O)[O-]
C6H3F2NO2
MFCD00012142
3,5-difluoronitrobenzene, benzene, 1,3-difluoro-5-nitro, 1,3-difluoro-5-nitro-benzene, 3,5-difluoro-1-nitrobenzene, 1-nitro-3,5-difluorobenzene, benzene, 1,3-difluoro-5-nitro-7ci,8ci,9ci, 3,5-difluoronitroben, pubchem2291, 3,5-difluornitrobenzene, acmc-1ck2x
1,3-difluoro-5-nitrobenzene

Specifications

74°C (165°F)
17°C
2265-94-3
MFCD00012142
AUQBBDWDLJSKMI-UHFFFAOYSA-N
1,3-difluoro-5-nitrobenzene
75272
98%
176°C to 177°C
1g
C6H3F2NO2
3,5-difluoronitrobenzene, benzene, 1,3-difluoro-5-nitro, 1,3-difluoro-5-nitro-benzene, 3,5-difluoro-1-nitrobenzene, 1-nitro-3,5-difluorobenzene, benzene, 1,3-difluoro-5-nitro-7ci,8ci,9ci, 3,5-difluoronitroben, pubchem2291, 3,5-difluornitrobenzene, acmc-1ck2x
C1=C(C=C(C=C1F)F)[N+](=O)[O-]
159.092
159.09
1,3-Difluoro-5-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CZ5712000