Please login to your online account to display your discounted pricing

1,3-Difluoro-2-propanol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Difluoro-2-propanol
Name Note 99%
CAS 453-13-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C3H6F2O
Linear Formula FCH2CH(OH)CH2F
Formula Weight 96.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC175560010

  SDS
thermo fisher scientific
AC175560010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear yellow to brownish
Physical Form Liquid
Formula Weight 96.08
Assay ≥97.5% (GC)
Quantity 1g
TSCA TSCA
RTECS Number UB1770000
Density 1.2400g/mL
EINECS Number 207-216-6
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 1 Warning
CAS Min % 98.5
Refractive Index 1.3720 to 1.3750 (20°C, 589nm)
Name Note 99%
CAS 453-13-4
MDL Number MFCD00000452
Chemical Name or Material 1, 3-Difluoro-2-propanol
Assay Percent Range 98%
Linear Formula FCH2CH(OH)CH2F
Molecular Formula C3H6F2O
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.