Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Difluoro-2-propanol, 98%

Chemical Identifiers

Chemical Name or Material 1,3-Difluoro-2-propanol
CAS 453-13-4
Molecular Formula C3H6F2O
Formula Weight 96.08
MDL Number MFCD00000452
UN Number UN1986
Beilstein 1732050
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0164603

  SDS
alfa aesar™
L0164603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0164606

  SDS
alfa aesar™
L0164606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 96.08
Chemical Name or Material 1,3-Difluoro-2-propanol
TSCA TSCA
Assay 98%
Molecular Formula C3H6F2O
MDL Number MFCD00000452
CAS 453-13-4
UN Number UN1986
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UB1770000
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, FLAMMABLE, TOXIC, N.O.S.
Flash Point 42°C (107°F)
Boiling Point 127°C
Quantity 1g
Beilstein 1732050
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.