Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Difluoro-2-propanol, 98%


Quantity 5g

Manufacturer: alfa aesar™  L0164606

 View more versions of this product

Catalog No. AAL0164606

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 96.08
Chemical Name or Material 1,3-Difluoro-2-propanol
CAS 453-13-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
MDL Number MFCD00000452
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Boiling Point 127°C
Quantity 5g
UN Number UN1986
Beilstein 1732050
Flash Point 42°C (107°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, FLAMMABLE, TOXIC, N.O.S.
Molecular Formula C3H6F2O
RTECS Number UB1770000

Specifications

Chemical Name or Material 1,3-Difluoro-2-propanol
CAS 453-13-4
Molecular Formula C3H6F2O
Formula Weight 96.08
MDL Number MFCD00000452
UN Number UN1986
Beilstein 1732050
Accessories

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents