Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Difluoro-2-propanol, 98%


Quantity 1g

Manufacturer: alfa aesar™  L0164603

 View more versions of this product

Catalog No. AAL0164603

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 96.08
Chemical Name or Material 1,3-Difluoro-2-propanol
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00000452
Health Hazard 1 Danger
Molecular Formula C3H6F2O
UN Number UN1986
RTECS Number UB1770000
CAS 453-13-4
Assay 98%
Health Hazard 2 GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Quantity 1g
Flash Point 42°C (107°F)
Boiling Point 127°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, FLAMMABLE, TOXIC, N.O.S.
Beilstein 1732050

Specifications

Chemical Name or Material 1,3-Difluoro-2-propanol
CAS 453-13-4
Molecular Formula C3H6F2O
Formula Weight 96.08
MDL Number MFCD00000452
UN Number UN1986
Beilstein 1732050
Accessories

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents