missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Difluoro-2-nitrobenzene, 98%

Manufacturer:  Alfa Aesar™ H55694MD

 View more versions of this product

Catalog No. AAH55694MD



Description

Description

Applications
1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
Specifications

Specifications

1,3-Difluoro-2-nitrobenzene
88°C (190°F)
91°C to 92°C (11mmHg)
250mg
C6H3F2NO2
1,3-difluoro-2-nitro-benzene
SSNCMIDZGFCTST-UHFFFAOYSA-N
1,3-difluoro-2-nitrobenzene
87922
98%
1.503
1.494
0°C
19064-24-5
MFCD00192035
Slightly soluble in water.
C1=CC(=C(C(=C1)F)[N+](=O)[O-])F
159.092
159.09

Chemical Identifiers

19064-24-5
159.092
SSNCMIDZGFCTST-UHFFFAOYSA-N
87922
C1=CC(=C(C(=C1)F)[N+](=O)[O-])F
C6H3F2NO2
MFCD00192035
1,3-difluoro-2-nitro-benzene
1,3-difluoro-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 242-793-8

TSCA : No

Recommended Storage : Ambient temperatures

Documents