missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Difluoro-2-nitrobenzene, 98%

Manufacturer:  Alfa Aesar™ H5569406

 View more versions of this product

Catalog No. AAH5569406



Description

Description

Applications
1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
Specifications

Specifications

1,3-Difluoro-2-nitrobenzene
91°C to 92°C (11mmHg)
5g
C6H3F2NO2
1,3-difluoro-2-nitro-benzene
C1=CC(=C(C(=C1)F)[N+](=O)[O-])F
159.092
159.09
88°C (190°F)
0°C
19064-24-5
MFCD00192035
SSNCMIDZGFCTST-UHFFFAOYSA-N
1,3-difluoro-2-nitrobenzene
87922
98%

Chemical Identifiers

19064-24-5
159.092
SSNCMIDZGFCTST-UHFFFAOYSA-N
87922
C1=CC(=C(C(=C1)F)[N+](=O)[O-])F
C6H3F2NO2
MFCD00192035
1,3-difluoro-2-nitro-benzene
1,3-difluoro-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents