missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Diethynylbenzene 96.0+%, TCI America™
$99.54 - $293.65
Chemical Identifiers
CAS | 1785-61-1 |
---|---|
Molecular Formula | C10H6 |
Molecular Weight (g/mol) | 126.158 |
MDL Number | MFCD00041686 |
InChI Key | PNXLPYYXCOXPBM-UHFFFAOYSA-N |
PubChem CID | 123130 |
IUPAC Name | 1,3-diethynylbenzene |
SMILES | C#CC1=CC(=CC=C1)C#C |
Chemical Identifiers
1785-61-1 | |
126.158 | |
PNXLPYYXCOXPBM-UHFFFAOYSA-N | |
1,3-diethynylbenzene |
C10H6 | |
MFCD00041686 | |
123130 | |
C#CC1=CC(=CC=C1)C#C |
Specifications
1785-61-1 | |
78°C | |
MFCD00041686 | |
PNXLPYYXCOXPBM-UHFFFAOYSA-N | |
1,3-diethynylbenzene | |
123130 | |
≥96.0% (GC) | |
1,3-Diethynylbenzene |
-3°C | |
C10H6 | |
1 g | |
C#CC1=CC(=CC=C1)C#C | |
126.158 | |
126.16 | |
Liquid |
Safety and Handling
TSCA : No