missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dicyanobenzene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 163660010

 View more versions of this product

Catalog No. AC163660010



Specifications

Specifications

1, 3-Dicyanobenzene
Authentic
288°C
161°C to 165°C
98%
97.5
97.5% min. (GC)
C6H4(CN)2
09, 836
Solubility in water: 0.7g/l (20°C). Other solubilities: 100g/l in acetone (20°C),insoluble in ether,acetone,hot ligroin,very soluble in alcohol,benzene and chloroform
C1=CC(=CC(=C1)C#N)C#N
128.13
CHEBI:38218
Flakes
>150°C
Plastic bottle
Tan to White
1kg
626-17-5
100.0
C8H4N2
MFCD00001795
1,3-benzenedicarbonitrile
LAQPNDIUHRHNCV-UHFFFAOYSA-N
benzene-1,3-dicarbonitrile
12276
128.13
98%

Chemical Identifiers

626-17-5
128.13
LAQPNDIUHRHNCV-UHFFFAOYSA-N
12276
benzene-1,3-dicarbonitrile
C8H4N2
MFCD00001795
1,3-benzenedicarbonitrile
CHEBI:38218
C1=CC(=CC(=C1)C#N)C#N
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-933-7

RTECSNumber : CZ1900000

TSCA : TSCA

SDS Documents