Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dichloropropene, cis + trans, tech. 90%

Chemical Identifiers

Chemical Name or Material 1,3-Dichloropropene
CAS 542-75-6
Name Note Cis + Trans
Purity Grade Technical Grade
Molecular Formula ClCH2CH=CHCl
Formula Weight 110.97
MDL Number MFCD00000985
UN Number UN2047
Beilstein 1719556
Merck Index 14,3075
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1033214

 
alfa aesar™
L1033214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1033222

 
alfa aesar™
L1033222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 110.97
Molecular Formula ClCH2CH=CHCl
MDL Number MFCD00000985
CAS 542-75-6
TSCA TSCA
Odor Chloroform-like
Name Note Cis + Trans
Purity Grade Technical Grade
Assay 90%
UN Number UN2047
Boiling Point 105° to 106°C
Beilstein 1719556
Melting Point ≈−84°C
Flash Point 21°C (70°F)
Merck Index 14,3075
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H311-H331-H350-H304-H341-H302-H315-H319-H317-H335
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
May be fatal if swallowed and enters airways.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DICHLOROPROPENES
RTECS Number UC8310000
Quantity 25g
Chemical Name or Material 1,3-Dichloropropene
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H331-H350-H304-H341-H302-H315-H319-H317-H335
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
May be fatal if swallowed and enters airways.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.