missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dichloropropene, cis + trans, tech. 90%

Manufacturer:  Alfa Aesar™ L1033222

 View more versions of this product

Catalog No. AAL1033222



Specifications

Specifications

1,3-Dichloropropene
Chloroform-like
~-84°C
Cis + Trans
C3H4Cl2
UN2047
14,3075
UOORRWUZONOOLO-OWOJBTEDSA-N
(E)-1,3-dichloroprop-1-ene
24726
110.97
Technical
21°C (70°F)
105°C to 106°C
100g
542-75-6
MFCD00000985
1719556
1,3-dichloropropene, trans-1,3-dichloropropene, telone ii, telone, 1,3-dichloropropylene, nemex, trans-1,3-dichloropropylene, dorlone ii, telone c, di-trapex cp
C(C=CCl)Cl
110.965
CHEBI:18624
90%

Chemical Identifiers

542-75-6
110.965
UOORRWUZONOOLO-OWOJBTEDSA-N
24726
(E)-1,3-dichloroprop-1-ene
C3H4Cl2
MFCD00000985
1,3-dichloropropene, trans-1,3-dichloropropene, telone ii, telone, 1,3-dichloropropylene, nemex, trans-1,3-dichloropropylene, dorlone ii, telone c, di-trapex cp
CHEBI:18624
C(C=CCl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H225-H311-H331-H350-H304-H341-H302-H315-H319-H317-H335
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
May be fatal if swallowed and enters airways.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DICHLOROPROPENES

RTECSNumber : UC8310000

TSCA : TSCA

Documents